Skip to Content
Merck
All Photos(1)

Key Documents

148547

4,4′-PCB

98%

Synonym(s):

4.4′-Dichlorobiphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8Cl2
CAS Number:
Molecular Weight:
223.10
EC Number:
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

98%

InChI

1S/C12H8Cl2/c13-11-5-1-9(2-6-11)10-3-7-12(14)8-4-10/h1-8H

InChI key

YTBRNEUEFCNVHC-UHFFFAOYSA-N

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Chu et al.
Water research, 36(9), 2187-2194 (2002-07-11)
Surfactant and organic solvents have individually been shown useful in assisting the solubilization of hydrophobic organics out of contaminated soil and promoting UV-induced photodecay at a succeeding treatment process. A newly proposed process is to use the mixtures of surfactant
L van Bree et al.
Toxicology, 42(2-3), 259-274 (1986-12-15)
Both 4,4'-dichlorobiphenyl (4,4'-DCB) and 3-methylcholanthrene (3-MC) caused a substantial increase of phenacetin-induced hepatic glutathione (GSH) depletion, whereas phenobarbital (PB) had no effect, suggesting that 4,4'-DCB possesses cytochrome P-448 inducing activity. The O-deethylation of phenacetin by liver microsomes from control and
Uptake of 4-chlorobiphenyl and 4,4'-dichlorobiphenyl in six species of plant tissue cultures.
K Yamamoto et al.
Bulletin of environmental contamination and toxicology, 28(6), 728-732 (1982-06-01)
P Adriaens et al.
Applied and environmental microbiology, 55(4), 887-892 (1989-04-01)
Acinetobacter sp. strain 4CB1 was isolated from a polychlorobiphenyl-contaminated soil sample by using 4-chlorobenzoate as a sole source of carbon and energy. Resting cells of Acinetobacter sp. strain 4CB1 hydrolytically dehalogenated 4-chlorobenzoate under aerobic and anaerobic conditions, but 4-hydroxybenzoate accumulated
D Dietrich et al.
Applied and environmental microbiology, 61(11), 3904-3909 (1995-11-01)
The white rot fungus Phanerochaete chrysosporium has demonstrated abilities to degrade many xenobiotic chemicals. In this study, the degradation of three model polychlorinated biphenyl (PCB) congeners (4,4'-dichlorobiphenyl [DCB], 3,3',4,4'-tetrachlorobiphenyl, and 2,2',4,4',5,5'-hexachlorobiphenyl) by P. chrysosporium in liquid culture was examined. After

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service