Skip to Content
Merck
All Photos(1)

Key Documents

14565

Sigma-Aldrich

α,ω-Bis{2-[(3-carboxy-1-oxopropyl)amino]ethyl}polyethylene glycol

Mr 2000

Synonym(s):

O,O′-Bis[2-(succinylamino)ethyl]polyethylene glycol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
R$3,474.00

R$3,474.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
1 G
R$3,474.00

About This Item

Linear Formula:
HOOCCH2CH2CONH(CH2CH2O)nCH2CH2NHCOCH2CH2COOH
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.22

R$3,474.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

mol wt

Mr 2000

SMILES string

O(COCCOCCC(=O)O)CCOCCC(=O)O

InChI

1S/C11H20O8/c12-10(13)1-3-16-5-7-18-9-19-8-6-17-4-2-11(14)15/h1-9H2,(H,12,13)(H,14,15)

InChI key

LQRCZNATFRANRS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service