Skip to Content
Merck
All Photos(1)

Documents

145521

Sigma-Aldrich

4-(Methylthio)benzoic acid

97%

Synonym(s):

4-(Methylmercapto)benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SC6H4CO2H
CAS Number:
Molecular Weight:
168.21
Beilstein:
2082809
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

192-196 °C (lit.)

SMILES string

CSc1ccc(cc1)C(O)=O

InChI

1S/C8H8O2S/c1-11-7-4-2-6(3-5-7)8(9)10/h2-5H,1H3,(H,9,10)

InChI key

KWHCPERWLHBLOT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Administration of 4-(methylthio)benzoic acid reduces cisplatin nephrotoxicity in rats. It prevents in vitro DNA binding and mutation induction in Escherichia coli K12.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P J Boogaard et al.
Biochemical pharmacology, 41(12), 1997-2003 (1991-06-15)
Administration of 4-methylthiobenzoic acid (MTBA) (100 mg/kg) strongly reduced cisplatin nephrotoxicity (7.5 mg/kg, 20 min after MTBA) in rats as determined by histopathology and blood urea nitrogen. Anti-tumour activity against a colonic adenocarcinoma, CC 531, that was implanted in rats
E D Kroese et al.
Mutation research, 245(2), 67-74 (1990-10-01)
Thioethers are effective scavengers of electrophilic metabolites derived from the hepatocarcinogen N-hydroxy-2-acetylaminofluorene (van den Goorbergh et al., 1987). In this study 2 of these thioethers, 4-(methylthio)benzoic acid (MTB) and its methylester, methyl 4-(methylthio)benzoate (MMTB), have been tested for their ability

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service