Skip to Content
Merck
All Photos(1)

Key Documents

139920

Sigma-Aldrich

8-Anilino-1-naphthalenesulfonic acid

97%

Synonym(s):

N-Phenyl peri acid, ANS, ANSA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHC10H6SO3H
CAS Number:
Molecular Weight:
299.34
Beilstein:
1843887
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

Assay

97%

impurities

<0.5% ethyl alcohol
<0.5% methyl alcohol
<1% acetone

mp

215-217 °C (dec.) (lit.)

SMILES string

OS(=O)(=O)c1cccc2cccc(Nc3ccccc3)c12

InChI

1S/C16H13NO3S/c18-21(19,20)15-11-5-7-12-6-4-10-14(16(12)15)17-13-8-2-1-3-9-13/h1-11,17H,(H,18,19,20)

InChI key

FWEOQOXTVHGIFQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Disruption of low- and high-density human plasma lipoproteins and phospholipid dispersions by 1-anilinonaphthalene-8-sulfonate.
R A Muesing et al.
Biochemistry, 10(15), 2952-2962 (1971-07-20)
Interaction of 8-amino-1-naphthalenesulfonate with rod outer segment membrane.
U P Andley et al.
Biochemistry, 20(6), 1687-1693 (1981-03-17)
Qing Zhang et al.
Carbohydrate polymers, 250, 116925-116925 (2020-10-15)
Influence of chitosan (Ch) with low, medium, and high molecular weight (LMW, MMW, and HMW) on the formation of field pea protein isolate (FPPI)/Ch complex coacervates was investigated. An increase in maximum turbidity and a gradual shift of critical pH
A C Saucier et al.
Biochemistry, 24(26), 7581-7585 (1985-12-17)
The binding of 8-anilino-1-naphthalenesulfonate (ANS) to ciliary dynein ATPase leads to a marked increase in the dye's fluorescence intensity, accompanied by a blue shift in the observed fluorescence emission maximum. We found that dynein has 37 +/- 3 ANS binding
Lauren E Schrock et al.
Movement disorders : official journal of the Movement Disorder Society, 30(4), 448-471 (2014-12-06)
Deep brain stimulation (DBS) may improve disabling tics in severely affected medication and behaviorally resistant Tourette syndrome (TS). Here we review all reported cases of TS DBS and provide updated recommendations for selection, assessment, and management of potential TS DBS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service