Skip to Content
Merck
All Photos(2)

Key Documents

138215

Sigma-Aldrich

2-Amino-4-methoxybenzothiazole

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8N2OS
CAS Number:
Molecular Weight:
180.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

mp

153-155 °C (lit.)

SMILES string

COc1cccc2sc(N)nc12

InChI

1S/C8H8N2OS/c1-11-5-3-2-4-6-7(5)10-8(9)12-6/h2-4H,1H3,(H2,9,10)

InChI key

YEBCRAVYUWNFQT-UHFFFAOYSA-N

Biochem/physiol Actions

2-Amino-4-methoxybenzothiazole on condensation reaction with 4-acetamidobenzaldehyde affords tridentate Schiff bases[1]. It reacts with 2,4,6-trichloro 1,3,5-triazine to give 2-(4-methoxybenzothiazol-2′-ylamino)-4-(phenylthioureido)-6-(substitutedthioureido)-1,3,5-triazines[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of Some New 2-(4-Methoxybenzothiazol-2'-yl amino)-4-(2-chloro-4-trifluoromethylanilino)-6-(substituted thioureido)-1, 3, 5-triazine as Antifungal Agents.
Sareen V, et al.
Phosph. Sulfur Relat. Elem., 185(1), 140-146 (2009)
Antibacterial Zn (II) compounds of. Schiff bases derived from some benzothiazoles.
Chohan ZH and Supuran CT.
Main Group Metal Chemistry, 25(5), 291-296 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service