Skip to Content
Merck
All Photos(1)

Key Documents

137472

Sigma-Aldrich

2,4-Dimethylanisole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3OCH3
CAS Number:
Molecular Weight:
136.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.514 (lit.)

bp

191 °C (lit.)

density

0.973 g/mL at 25 °C (lit.)

SMILES string

COc1ccc(C)cc1C

InChI

1S/C9H12O/c1-7-4-5-9(10-3)8(2)6-7/h4-6H,1-3H3

InChI key

UJCFZCTTZWHRNL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,4-Dimethylanisole was used to study the solvolysis of 1,4-dimethyl-4-nitrocyclohexa-2,5-dien-1. It was used to study the photolysis of charge transfer complex of 2,4-dimethylanisole with tetranitromethane.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photochemical Nitration by Tetranitromethane. Part XXXVII. t Adduct Formation and the Regiochemistry of Attack of Trinitromethanide Ion on Radical Cations in the Photochemical Reactions of 2-Methyl-, 2, 3-Dimethyl-and 2, 4-Dimethylanisoles.
Timmerman-Vaughan DJ.
Acta Chemica Scandinavica, 51(19971), 73-87 (1997)
The role of ion-molecule pair intermediates in acid-catalyzed solvolysis. General base-catalyzed formation of 4-methylbenzyl carbocation and its trapping by nucleophiles.
Thibblin A.
The Journal of Organic Chemistry, 58(26), 7427-7433 (1993)
Reactions of the 1-hydroxy-1, 4-dimethylcyclohexadienyl cation, an intermediate in the solvolysis of 1, 4-dimethyl-4-nitrocyclohexa-2, 5-dien-1-ol.
Fischer A, et al.
Canadian Journal of Chemistry, 64(6), 1093-1101 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service