Skip to Content
Merck
All Photos(2)

Key Documents

134376

Sigma-Aldrich

Succinamic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCOCH2CH2CO2H
CAS Number:
Molecular Weight:
117.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
97%

Assay

97%

mp

153-156 °C (lit.)

functional group

amide
carboxylic acid

SMILES string

NC(=O)CCC(O)=O

InChI

1S/C4H7NO3/c5-3(6)1-2-4(7)8/h1-2H2,(H2,5,6)(H,7,8)

InChI key

JDVPQXZIJDEHAN-UHFFFAOYSA-N

General description

Succinamic acid forms binuclear europium complexes. It forms mixed valent platinum blues with K2PtCl4.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure and photophysical properties of europium complexes of succinamic acid and 1, 10-phenanthroline.
Zheng X-J, et al.
Polyhedron, 22(2), 323-330 (2003)
Synthesis, binding sites, and spectroscopic characterizations of succinamic acid and succinimide platinum blues.
Arrizabalaga P, et al.
Journal of the American Chemical Society, 106(5), 1300-1303 (1984)
Vincent Michel et al.
Scientific reports, 10(1), 16430-16430 (2020-10-04)
The hair bundle of cochlear hair cells is the site of auditory mechanoelectrical transduction. It is formed by three rows of stiff microvilli-like protrusions of graduated heights, the short, middle-sized, and tall stereocilia. In developing and mature sensory hair cells
Yasuhiko Kizuka et al.
Cell chemical biology, 24(12), 1467-1478 (2017-10-17)
Fucosylation is a glycan modification critically involved in cancer and inflammation. Although potent fucosylation inhibitors are useful for basic and clinical research, only a few inhibitors have been developed. Here, we focus on a fucose analog with an alkyne group
Ye Wang et al.
Frontiers in microbiology, 8, 435-435 (2017-04-04)
Commensal non-toxigenic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service