Skip to Content
Merck
All Photos(3)

Key Documents

132713

Sigma-Aldrich

2,6-Dimethyl-4-nitrophenol

98%

Synonym(s):

4-Nitro-2,6-xylenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H2(CH3)2OH
CAS Number:
Molecular Weight:
167.16
Beilstein:
1873275
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

168 °C (dec.) (lit.)

functional group

nitro

SMILES string

Cc1cc(cc(C)c1O)[N+]([O-])=O

InChI

1S/C8H9NO3/c1-5-3-7(9(11)12)4-6(2)8(5)10/h3-4,10H,1-2H3

InChI key

FNORUNUDZNWQFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,6-Dimethyl-4-nitrophenol was used to study the disposition requirement for binding of p-nitrophenol and sodium p-nitrophenolate like substrates with cyclohexaamylaose[1]. It was used as internal standard in determination of 4-nitrophenol and 3-methyl-4-nitrophenol in human urine by liquid chromatography combined with tandem mass spectrometry [2].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan V Sancho et al.
Rapid communications in mass spectrometry : RCM, 16(7), 639-645 (2002-03-29)
The potential of liquid chromatography combined with tandem mass spectrometry (LC/MS/MS) for the determination of pesticide metabolites in human urine at the sub-ppb level is explored. Metabolites from two organophosphorous pesticides, 4-nitrophenol (from parathion and parathion-methyl) and 3-methyl-4-nitrophenol (from fenitrothion)
Disposition requirements for binding in aqueous solution of polar substrates in the cyclohexaamylose cavity.
Bergeron RJ, et al.
Journal of the American Chemical Society, 99(15), 5146-5151 (1977)
Tomohiro Ito et al.
Environmental science and pollution research international, 26(22), 22747-22755 (2019-06-07)
Secondary organic aerosol (SOA) is a component of airborne particulate matter in urban areas. However, their toxicities remain incompletely understood. In this study, we investigated the oxidative and inflammatory potency of SOA derived from three different volatile organic compounds (α-pinene

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service