Skip to Content
Merck
All Photos(1)

Key Documents

124206

Sigma-Aldrich

4-Butoxybenzoic acid

98%

Synonym(s):

p -(Butyloxy)benzoic acid, p -Butoxybenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3OC6H4CO2H
CAS Number:
Molecular Weight:
194.23
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

liquid crystal

mp

147-150 °C (lit.)

SMILES string

CCCCOc1ccc(cc1)C(O)=O

InChI

1S/C11H14O3/c1-2-3-8-14-10-6-4-9(5-7-10)11(12)13/h4-7H,2-3,8H2,1H3,(H,12,13)

InChI key

LAUFPZPAKULAGB-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, liquid crystalline properties and thermal stability of 4-(4-alkyloxyphenylazo) benzoic acids
Cioanca, Elena-Raluca, et al.
Analele Universitatii Bucuresti: Chimie , 19(2) (2010)
Induced crystal G Phase through intermolecular hydrogen bonding in nOBA: 9HB
S. Sriharisastry, C.Nageswara Rao, T. Vishwam, K. Mallika, B. Gowrishankar Rao, Ha Sie Tiong.
Research Journal of Physical Sciences, 1(6), 1-10 (2013)
Rama Krishna Gudiminchi et al.
Applied microbiology and biotechnology, 90(1), 117-126 (2010-12-17)
CYP102A1 is an efficient medium- to long-chain fatty acid hydroxylase that is able to accept a wide range of non-natural substrates which bear no resemblance to the natural ones. 4-Hexylbenzoic acid (HBA) and 4-nonyloxybenzoic acid (NOBA) were identified as CYP102A1
Rheological study of structural transitions in triblock copolymers in a liquid crystal solvent.
Kempe, Michael D., et al.
Soft Matter, 2(5), 422-431 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service