Skip to Content
Merck
All Photos(1)

Key Documents

123358

Sigma-Aldrich

1-Chloro-2-methylpropene

98%

Synonym(s):

Isocrotyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCl
CAS Number:
Molecular Weight:
90.55
Beilstein:
1733843
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.424 (lit.)

bp

68 °C (lit.)

density

0.92 g/mL at 25 °C (lit.)

functional group

alkyl halide
chloro

SMILES string

C\C(C)=C\Cl

InChI

1S/C4H7Cl/c1-4(2)3-5/h3H,1-2H3

InChI key

KWISWUFGPUHDRY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Chloro-2-methylpropene undergoes hydroboration to form an adduct, which on oxidation forms isobutyraldehyde.

Application

1-Chloro-2-methylpropene was used to study multiple infrared photon excitation of α-chloro olefins.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Dermal - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

30.2 °F - closed cup

Flash Point(C)

-1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R E Cannon et al.
Molecular carcinogenesis, 29(4), 229-235 (2001-02-15)
This work was initiated to determine the potential for the Tg.AC mouse model to identify chemical carcinogens by an oral route of administration. Tg.AC v-Ha-ras transgenic mice were exposed to dimethyvinyl chloride (DMVC; 1-chloro-2-methylpropene), a structural analog of the human
Formation of vinylidenecarbene intermediates in multiple infrared photon elimination reactions.
Reiser C and Steinfeld JI.
The Journal of Physical Chemistry, 84(6), 680-681 (1980)
Dimethylvinyl chloride.
Report on carcinogens : carcinogen profiles, 11, III110-III110 (2004-01-01)
1-Chloro-2-methylpropene.
IARC monographs on the evaluation of carcinogenic risks to humans, 63, 315-324 (1995-01-01)
P Srinivas et al.
Drug metabolism and disposition: the biological fate of chemicals, 16(3), 449-454 (1988-05-01)
Recent metabolism and disposition studies of 1-chloro-2-methylpropene (dimethylvinyl chloride) revealed cysteine and N-acetylcysteine conjugates of 3-chloro-2-methylpropenoic acid as the major metabolites. In the present studies we have investigated various steps in the metabolic pathway to determine which step was responsible

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service