Skip to Content
Merck
All Photos(1)

Key Documents

118133

Sigma-Aldrich

2-Cyanophenylacetonitrile

97%

Synonym(s):

α-Cyano-o-tolunitrile, Homophthalonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
R$670.00

R$670.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
R$670.00

About This Item

Linear Formula:
NCC6H4CH2CN
CAS Number:
Molecular Weight:
142.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$670.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

functional group

nitrile

SMILES string

N#CCc1ccccc1C#N

InChI

1S/C9H6N2/c10-6-5-8-3-1-2-4-9(8)7-11/h1-4H,5H2

InChI key

GKHSEDFDYXZGCG-UHFFFAOYSA-N

General description

2-Cyanophenylacetonitrile undergoes carbocyclization reaction with bromoacrylonitriles to give naphthylamine[1].

Application

2-Cyanophenylacetonitrile may be used in the synthesis of 2, 5-diphenylpyrazine[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Single step synthesis of 2, 4-dicyanonaphthylamines from synthetic equivalents of a-cyanoalkynes.
Reux D and Pochat F.
Journal of the Chemical Society. Chemical Communications, 20, 1419-1420 (1991)
A novel synthesis of 2, 5-diphenylpyrazine.
Luo M, et al.
Research on Chemical Intermediates, 36(5), 491-494 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service