Skip to Content
Merck
All Photos(1)

Documents

11629

Sigma-Aldrich

Diethyl azodicarboxylate on polystyrene

100-300 mesh, extent of labeling: ~1.2 mmol/g N loading

Synonym(s):

DEAD polymer-bound, Polymer DEAD

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: solution phase peptide synthesis

extent of labeling

~1.2 mmol/g N loading

matrix

crosslinked with 1% DVB

particle size

100-300 mesh

storage temp.

2-8°C

Other Notes

Substitute for soluble DEAD in Mitsunobu reactions which is easily separable and non-explosive

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

L.D. Arnold et al.
Journal of the American Chemical Society, 111, 3973 -3973 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service