Skip to Content
Merck
All Photos(1)

Documents

110124

Sigma-Aldrich

1,2,3-Trichloropropane

99%

Synonym(s):

Glycerol trichlorohydrin, Trichlorohydrin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2ClCHClCH2Cl
CAS Number:
Molecular Weight:
147.43
Beilstein:
1732068
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39050132
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.484 (lit.)

bp

156 °C (lit.)

mp

−14 °C (lit.)

density

1.387 g/mL at 25 °C (lit.)

SMILES string

ClCC(Cl)CCl

InChI

1S/C3H5Cl3/c4-1-3(6)2-5/h3H,1-2H2

InChI key

CFXQEHVMCRXUSD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2,3-Trichloropropane is reductively dehalogenated by the anaerobic bacterial strain within chloroflexi.

Application

1,2,3-trichloropropane has been used to study its reaction with OH regarding it spotential atmospheric relevance.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Repr. 1B - STOT RE 1 Inhalation

Target Organs

Kidney,Liver,Mucous membranes

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rate constants for the reactions of OH with chlorinated propanes.
Yujing M and Mellouki A.
Physical Chemistry Chemical Physics, 3(13), 2614-2617 (2001)
Ghufrana Samin et al.
Environmental science and pollution research international, 19(8), 3067-3078 (2012-08-10)
1,2,3-Trichloropropane (TCP) is a persistent groundwater pollutant and a suspected human carcinogen. It is also is an industrial chemical waste that has been formed in large amounts during epichlorohydrin manufacture. In view of the spread of TCP via groundwater and
Eakalak Khan et al.
Journal of hazardous materials, 161(2-3), 1024-1034 (2008-05-27)
The mineralization and biodegradability increase and their combination of two traditional and two relatively new organic contaminants by Fenton reagents with three different types of iron, Fe(2+), Fe(3+), and Fe(0) were investigated. The traditional contaminants examined were trichloroethene (TCE) and
Vaishnavi Sarathy et al.
Environmental science & technology, 44(2), 787-793 (2009-12-17)
1,2,3-Trichloropropane (TCP) is an emerging contaminant because of increased recognition of its occurrence in groundwater, potential carcinogenicity, and resistance to natural attenuation. The physical and chemical properties of TCP make it difficult to remediate, with all conventional options being relatively
Grace E Kissling et al.
Toxicological sciences : an official journal of the Society of Toxicology, 92(1), 143-156 (2006-04-04)
There has been considerable interest in the use of small fish models for detecting potential environmental carcinogens. In this study, both guppies (Poecilia reticulata) and medaka (Oryzias latipes) were exposed in the aquaria water to three known rodent carcinogens for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service