Skip to Content
Merck
All Photos(1)

Key Documents

110000

Sigma-Aldrich

Diethylamine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NH
CAS Number:
Molecular Weight:
73.14
Beilstein:
605268
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
technique(s):
ELISA: suitable
bp:
55 °C (lit.)
vapor pressure:
14.14 psi ( 55 °C)
180 mmHg ( 20 °C)
3.51 psi ( 20 °C)
Pricing and availability is not currently available.

vapor density

2.5 (vs air)

vapor pressure

14.14 psi ( 55 °C)
180 mmHg ( 20 °C)
3.51 psi ( 20 °C)

Assay

98%

form

liquid

autoignition temp.

594 °F

expl. lim.

10.1 %

technique(s)

ELISA: suitable

refractive index

n20/D 1.385 (lit.)

pH

13 (20 °C, 100 g/L)

bp

55 °C (lit.)

mp

−50 °C (lit.)

density

0.707 g/mL at 25 °C (lit.)

SMILES string

CCNCC

InChI

1S/C4H11N/c1-3-5-4-2/h5H,3-4H2,1-2H3

InChI key

HPNMFZURTQLUMO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-14.8 °F - closed cup

Flash Point(C)

-26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Guo et al.
Journal of chromatography. A, 1307, 66-72 (2013-08-15)
The extraction efficiency of analytes in a cold fiber headspace solid-phase microextraction (CF-HS-SPME) system was investigated both theoretically and experimentally. The system was applied for quantitative extraction of polycyclic aromatic hydrocarbons (PAHs) from solid matrices. In order to achieve better
Rakesh Naduvile Veedu et al.
The journal of physical chemistry. A, 112(40), 9742-9750 (2008-09-16)
Five aryliminopropadienones 4a- d have been synthesized by flash vacuum thermolysis (FVT) by using two different precursors in each case. These compounds were deposited at 50 K at a pressure of ca. 10(-6) mbar together with three different nucleophiles, namely
Doron Yariv et al.
Colloids and surfaces. B, Biointerfaces, 78(2), 185-192 (2010-04-07)
In this paper we examined feasible correlations between the structure of different lyotropic mesophases and transdermal administration of three diclofenac derivatives with varying degrees of kosmotropic or chaotropic properties, solubilized within the mesophases. It was found that the most chaotropic
Katina S S Dossou et al.
Journal of chromatography. A, 1216(44), 7450-7455 (2009-06-26)
A polysaccharide-based chiral stationary phase (Sepapak-4), with cellulose tris(4-chloro-3-methylphenylcarbamate) as chiral selector, has been investigated in liquid chromatography (LC). Its enantioresolution power was evaluated towards 13 basic amino-drugs with widely different structures and polarities, using polar organic mobile phases. After
Roberto Cirilli et al.
Analytical chemistry, 81(9), 3560-3570 (2009-03-28)
Second-order rate constants of the diethylamine-promoted enantiomerization of 2-[2-(1-methyl-1H-pyrrol-2-yl)-2-oxo-1-phenylethyl]-isoindole-1,3-dione, a chiral alpha-substituted ketone endowed with high anti-MAO activity type-A, were measured by dynamic high-performance liquid chromatography (DHPLC), stopped-flow high-performance liquid chromatography (sf-HPLC), and a classical method based on enantioselective HPLC

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service