Skip to Content
Merck
All Photos(1)

Key Documents

102180

Sigma-Aldrich

1-Cyanoacetylpiperidine

98%

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
R$3,489.00

R$3,489.00


Estimated to ship onApril 17, 2025


Request a Bulk Order

Select a Size

Change View
100 G
R$3,489.00

About This Item

Empirical Formula (Hill Notation):
C8H12N2O
CAS Number:
Molecular Weight:
152.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$3,489.00


Estimated to ship onApril 17, 2025


Request a Bulk Order

Quality Level

Assay

98%

form

solid

mp

88-91 °C (lit.)

SMILES string

O=C(CC#N)N1CCCCC1

InChI

1S/C8H12N2O/c9-5-4-8(11)10-6-2-1-3-7-10/h1-4,6-7H2

InChI key

ANLQHFYDQPMDJY-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service