Skip to Content
Merck
All Photos(2)

Key Documents

02615

Sigma-Aldrich

(Ethoxycarbonylmethyl)triphenylphosphonium chloride

technical, ≥90% (AT)

Synonym(s):

(2-Ethoxy-2-oxoethyl)triphenylphosphonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OCOCH2P(Cl)(C6H5)3
CAS Number:
Molecular Weight:
384.84
Beilstein:
3924139
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

≥90% (AT)

reaction suitability

reaction type: C-C Bond Formation

mp

120-123 °C (dec.)
120-123 °C (lit.)

functional group

ester
phosphine

storage temp.

2-8°C

SMILES string

[Cl-].CCOC(=O)C[P+](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C22H22O2P.ClH/c1-2-24-22(23)18-25(19-12-6-3-7-13-19,20-14-8-4-9-15-20)21-16-10-5-11-17-21;/h3-17H,2,18H2,1H3;1H/q+1;/p-1

InChI key

DJGHVEPNEJKZBF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Preparation of disubstituted tetrahydrofurans as selective serotonin re-uptake inhibitors for the treatment of depression
  • Ring-opening reactions with Wittig reagents
  • Enantioselective total synthesis of the phytotoxic lactone herbarumin III via Keck′s asymmetric allylation and Sharpless epoxidation
  • Synthesis of fluorobenzofurans via microwave-assisted tandem intramolecular Wittig and Claisen rearrangement reactions
  • Preparation of phosphonium derivatives of coumarin

Other Notes

Starting material for preparing the Wittig-reagent[1][2][3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

H.J. Bestmann et al.
Chemische Berichte, 96, 465-465 (1963)
O. Sieber et al.
Helvetica Chimica Acta, 40, 1242-1242 (1957)
H. Moison et al.
Tetrahedron, 43, 537-537 (1987)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service