Skip to Content
Merck
All Photos(1)

Documents

AC7300012537

Advanced Chemical Intermediates Ltd.

N'-[2-(4-Chlorophenyl)-1-cyclopropylaminoethylidene]hydrazinecarboxylic acid tert-butyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
323.82
MDL number:

Iupac Name

tert-butyl (2E)-2-[2-(4-chlorophenyl)-1-(cyclopropylamino)ethylidene]hydrazinecarboxylate

Inchi Code

1S/C16H22ClN3O2/c1-16(2,3)22-15(21)20-19-14(18-13-8-9-13)10-11-4-6-12(17)7-5-11/h4-7,10,13,18-19H,8-9H2,1-3H3,(H,20,21)

InChI key

IZWLLFTXJUMMMA-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

95%

Country of Origin

GB

Shipping Temperature

Room temperature

Physical Form

beige solid

Product Link

https://www.acints.com/products/ACI-02A-040

Pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.acints.com/products/ACI-02A-040


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a productā€™s label following the words ā€˜Lotā€™ or ā€˜Batchā€™.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service