Skip to Content
Merck
All Photos(1)

Documents

ADVH9454742D

AChemBlock

ethyl 4,5,6,7-tetrahydrothieno[2,3-c]pyridine-2-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
211.28
MDL number:

Iupac Name

ethyl 4,5,6,7-tetrahydrothieno[2,3-c]pyridine-2-carboxylate

Inchi Code

1S/C10H13NO2S/c1-2-13-10(12)8-5-7-3-4-11-6-9(7)14-8/h5,11H,2-4,6H2,1H3

InChI key

KEZGGJBGQAHGRQ-UHFFFAOYSA-N

storage temp.

Store at 0-8 C

Purity

95%

Country of Origin

CN

Product Link

https://www.achemblock.com/t93176-ethyl-4-5-6-7-tetrahydrothieno-2-3-c-pyridine-2-carboxylate.html

Synonym

ethyl 4,5,6,7-tetrahydrothieno[2,3-c]pyridine-2-carboxylate

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://www.achemblock.com/t93176-ethyl-4-5-6-7-tetrahydrothieno-2-3-c-pyridine-2-carboxylate.html


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service