Merck
mAbs Downstream
Products for Development and Manufacturing
Higher cell densities and antibody titers from increasingly advanced upstream processes result in complex downstream purification challenges. The focus of downstream bioprocessing is efficient recovery and purification while controlling bioburden and assuring viral safety to provide drug safety for patients. The downstream workflow, which includes clarification, chromatography, concentration, viral clearance, and sterile filtration steps, is essential for ensuring monoclonal antibody (mAb) product quality, yield, safety, and sterility. 
Clarification
Clarification, both primary and secondary, is used to separate secreted monoclonal antibodies (mAbs) from whole cells, colloids, aggregated and non-product proteins, lipids, and large particles in the cell culture fluid harvest. More than one clarification step is typically required for removal of these impurities and to prepare the feedstream for further downstream purification. Various types of pre-treatments (e.g., precipitation/flocculation) are increasingly employed as effective methods to clarify high density cell cultures and to make the clarification process more efficient. Centrifugation, depth filtration, and sterile filtration are the most common approaches to clarify mammalian cell harvest material and effectively recover antibody for downstream processing.
Our comprehensive offering for harvest clarification in monoclonal antibody (mAb) development and production processes includes:
  • Millipore® Filters (clarification depth filters & pretreatment solutions, prefilters)
  • Millipore® / SAFC® Buffer and Media Preparation
  • Millipore® Single-Use Systems and Filter Holders
  • SAFC® Buffers and CIP Solutions
  • Millipore® Services (single-use and filter validation, equipment installation, system qualification, system training)
  • Millipore® Automation and Analytics Software
Related Products
Loading
Related Categories
Four containers are depicted in the image related to buffers and pH adjusters. The two smaller containers on the left have a red lid and white body. The two larger containers on the right are blue with red-rimmed lids.
Buffers & pH Adjusters
Our broad portfolio of buffer materials manufactured under appropriate controls is tailored to your needs.
Learn More
An industrial clarification filter system featuring multiple cylindrical units mounted on a metal frame, complete with gauges and valves, designed for large-scale filtration processes.
Clarification & Harvest
Clarification steps prepare your bioreactor harvest material for downstream purification by reducing the levels of impurities and particulates.
Learn More
Chromatograph system for bioprocessing, featuring various tubes, dials, and components, predominantly silver with blue accents and a logo featuring the letter ‘M’ on the side panel.
Chromatography
Downstream purification in bioprocessing includes one or more chromatographic separation steps to purify your molecule and remove impurities.
Learn More
A female scientist wearing protective goggles holds up a sterile drug product.
Sterile Filtration and Bioburden Control
Our broad portfolio of membrane filters offers options for efficient processing of every pharma and biopharma stream.
Learn More
Dropper transferring liquid in monoclonal antibody process
Viral Clearance
Our viral clearance portfolio includes the virus filtration solutions, chemicals, holders and systems.
Learn More
Mobius® FlexReady Solution for TFF
Ultrafiltration & Diafiltration
Ultrafiltration & diafiltration increase capacity, concentration, and recovery in bioprocessing.
Learn More
Related Resources
Explore Our Solutions