Merck
  • Allogeneic bone marrow mesenchymal stem cell transplantation in patients with UDCA-resistant primary biliary cirrhosis.
Allogeneic bone marrow mesenchymal stem cell transplantation in patients with UDCA-resistant primary biliary cirrhosis.
Stem cells and development (2014-05-20)
Li Wang, Qin Han, Hua Chen, Ke Wang, Guang-liang Shan, Fang Kong, Yun-jiao Yang, Yong-zhe Li, Xuan Zhang, Fen Dong, Qian Wang, Dong Xu, Zhao-jun Hu, Shi-hua Wang, Armand Keating, Ya-lan Bi, Feng-chun Zhang, Robert Chun-hua Zhao
ABSTRACT
The objective of this study was to evaluate the safety and efficacy of allogeneic bone marrow mesenchymal stromal/stem cell transplantation (BM-MSCT) for patients with ursodeoxycholic acid (UDCA)-resistant primary biliary cirrhosis (PBC). Ten patients were enrolled in this trial of BM-MSCT. All patients were permitted to concurrently continue their previous UDCA treatment. The efficacy of BM-MSCT in UDCA-resistant PBC was assessed at various time points throughout the 12-month follow up. No transplantation-related side effects were observed. The life quality of the patients was improved after BM-MSCT as demonstrated by responses to the PBC-40 questionnaire. Serum levels of ALT, AST, γ-GT, and IgM significantly decreased from baseline after BM-MSCT. In addition, the percentage of CD8+ T cells was reduced, while that of CD4+CD25+Foxp3+ T cells was increased in peripheral lymphocytic subsets. Serum levels of IL-10 were also elevated. Notably, the optimal therapeutic outcome was acquired in 3 to 6 months and could be maintained for 12 months after BM-MSCT. In conclusion, allogeneic BM-MSCT in UDCA-resistant PBC is safe and appears to be effective.
Product Description
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Selenium, pellets, < 5mm, ≥99.999%
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethasone, VETRANAL®, analytical standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Dexamethasone, British Pharmacopoeia (BP) Assay Standard