Merck
  • Mosquito larvicidal activity of methyl-p-hydroxybenzoate isolated from the leaves of Vitex trifolia Linn.
Mosquito larvicidal activity of methyl-p-hydroxybenzoate isolated from the leaves of Vitex trifolia Linn.
Acta tropica (2011-07-19)
K Kannathasan, A Senthilkumar, V Venkatesalu
ABSTRACT
The vector-borne diseases caused by mosquitoes are one of the major health problems in many countries especially in tropical and sub-tropical countries. The resistance of mosquitoes to synthetic chemicals and environmental toxicity created by the chemicals raised the demand for finding of alternate natural molecules that control mosquito. In the present study, a crystalline compound methyl-p-hydroxybenzoate was isolated from the methanol extract of Vitex trifolia leaves and it was identified by (1)H and (13)C NMR and single crystal X-ray diffractometer. The larvicidal potential of the isolated compound was evaluated against early 4th instar larvae of Culex quinquefasciatus and Aedes aegypti. The compound exhibited 100% larval mortality of both the mosquitoes at 20 ppm with LC(50) values of 5.77 and 4.74 ppm against C. quinquefasciatus and A. aegypti, respectively. The methyl-p-hydroxybenzoate, which is reported for the first time to our best of knowledge from V. trifolia can be better explored for the control of mosquito population.
Sigma-Aldrich
Methyl 4-hydroxybenzoate, analytical standard
Sigma-Aldrich
Methyl 4-hydroxybenzoate, tested according to Ph. Eur.
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ≥99%, FCC
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ReagentPlus®, ≥99.0%, crystalline
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Methyl 4-hydroxybenzoate, SAJ first grade, ≥98.0%
Supelco
Methyl Paraben, PESTANAL®, analytical standard
Methyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
Supelco
Methylparaben, Pharmaceutical Secondary Standard; Certified Reference Material