Merck
  • The addition of lysostaphin dramatically improves survival, protects porcine biomesh from infection, and improves graft tensile shear strength.
The addition of lysostaphin dramatically improves survival, protects porcine biomesh from infection, and improves graft tensile shear strength.
The Journal of surgical research (2011-06-24)
Igor Belyansky, Victor B Tsirline, Terri R Martin, David A Klima, Jessica Heath, Amy E Lincourt, Rohan Satishkumar, Alexey Vertegel, B Todd Heniford
ABSTRACT
Lysostaphin (LS), a naturally occurring Staphylococcal endopeptidase, has the ability to penetrate biofilm, and has been identified as a potential antimicrobial to prevent mesh infection. The goals of this study were to determine if LS adhered to porcine mesh (PM) can impact host survival, reduce the risk of long-term PM infection, and to analyze lysostaphin bound PM (LS-PM) mesh-fascial interface in an infected field. Abdominal onlay PMs measuring 3×3 cm were implanted in select groups of rats (n=75). Group assignments were based on bacterial inoculum and presence of LS on mesh. Explantation occurred at 60 d. Bacterial growth and mesh-fascial interface tensile strength were analyzed. Standard statistical analysis was performed. Only one out of 30 rats with bacterial inoculum not treated with LS survived. All 30 LS treated rats survived and had normal appearing mesh, including 20 rats with a bacterial inoculum (10(6) and 10(8) CFU). Mean tensile strength for controls and LS and no inoculum samples was 3.47±0.86 N versus 5.0±1.0 N (P=0.008). LS groups inoculated with 10(6) and 10(8) CFU exhibited mean tensile strengths of 4.9±1.5 N and 6.7±1.6 N, respectively (P=0.019 and P<0.001 compared with controls). Rats inoculated with S. aureus and not treated with LS had a mortality of 97%. By comparison, LS treated animals completely cleared S. aureus when challenged with bacterial concentrations of 1×10(6) and 1×10(8) with maintenance of mesh integrity at 60 d. These findings strongly suggest the clinical use of LS-treated porcine mesh in contaminated fields may translate into more durable hernia repair.
Sigma-Aldrich
Lysostaphin from Staphylococcus simulans, recombinant, expressed in E. coli, lyophilized powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, lyophilized powder, Protein 50-70 % by biuret, ≥500 units/mg protein
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, aseptically filled
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, BioUltra, ≥97% (SDS-PAGE), Protein 40-60 % by biuret, ≥2,000 units/mg protein