
mPEG5K

Synonym(s):
mPEG-Silane, mPEG-Alkyne, mPEG-Propionaldehyde, mPEG-Hydrazide
Linear Formula:
CH3O(CH2CH2O)nCH2CONHNH2
Products
Brand
Physical Form
Functionality
Shape
Polymer Composition
Polymer Type
Reactivity
Reagent Type
Showing 1-10 of 10
Compare
Product Number
Description
Pricing