
Acetaminophen Impurity I

Synonym(s):
2′-Hydroxyacetophenone, 2-Acetylphenol
Linear Formula:
HOC6H4COCH3
CAS Number:
Molecular Weight:
136.15
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Format
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing