PHR1973
Acetaminophen Impurity I
Pharmaceutical Secondary Standards; Certified Reference Material
Synonym(s):
2′-Hydroxyacetophenone, 2-Acetylphenol
About This Item
grade
certified reference material
pharmaceutical secondary standard
Quality Level
vapor density
4.7 (vs air)
vapor pressure
~0.2 mmHg ( 20 °C)
CofA
current certificate can be downloaded
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
refractive index
n20/D 1.558 (lit.)
bp
213 °C/717 mmHg (lit.)
mp
3-6 °C (lit.)
density
1.133 g/mL at 20 °C
1.131 g/mL at 25 °C (lit.)
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-30°C
SMILES string
CC(=O)c1ccccc1O
InChI
1S/C8H8O2/c1-6(9)7-4-2-3-5-8(7)10/h2-5,10H,1H3
InChI key
JECYUBVRTQDVAT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Analysis Note
Other Notes
Footnote
Recommended products
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
222.8 °F - closed cup
Flash Point(C)
106 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service