Skip to Content
Merck
All Photos(1)

Key Documents

1A00930

USP

Methyl p-toluate

new

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Methyl p-toluate, (Methyl 4-methylbenzoate)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CO2CH3
CAS Number:
Molecular Weight:
150.17
Beilstein:
1100609
MDL number:

grade

pharmaceutical primary standard

Agency

USP

vapor density

4.7 (vs air)

vapor pressure

1 mmHg ( 39 °C)

autoignition temp.

932 °F

expl. lim.

9.5 %

manufacturer/tradename

USP

bp

103-104 °C/15 mmHg (lit.)

mp

32-35 °C (lit.)

application(s)

pharmaceutical small molecule

format

neat

storage temp.

-10 to -25°C

SMILES string

COC(=O)c1ccc(C)cc1

InChI

1S/C9H10O2/c1-7-3-5-8(6-4-7)9(10)11-2/h3-6H,1-2H3

InChI key

QSSJZLPUHJDYKF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

USP issued SDS can be found here
Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service