Skip to Content
Merck

1670207

USP

Tolcapone

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

3,4-Dihydroxy-4′-methyl-5-nitrobenzophenone, Ro 40-7592

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H11NO5
CAS Number:
Molecular Weight:
273.24
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

tolcapone

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cc1ccc(cc1)C(=O)c2cc(O)c(O)c(c2)[N+]([O-])=O

InChI

1S/C14H11NO5/c1-8-2-4-9(5-3-8)13(17)10-6-11(15(19)20)14(18)12(16)7-10/h2-7,16,18H,1H3

InChI key

MIQPIUSUKVNLNT-UHFFFAOYSA-N

Gene Information

human ... COMT(1312)

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Tolcapone USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Tolcapone Tablets

Biochem/physiol Actions

Orally active catechol-O-methyltransferase (COMT) inhibitor: inhibits both central and peripheral COMT.
Tolcapone is an orally active catechol-O-methyltransferase (COMT) inhibitor. It inhibits both central and peripheral COMT. Tolcapone crosses the blood-brain barrier, and has been used for L-DOPA adjunct therapy in the treatment of Parkinson′s Disease.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Still not finding the right product?

Explore all of our products under Tolcapone


pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library