1272907
USP
Fludeoxyglucose
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
2-Fluoro-2-deoxy-D-glucose, 2-Deoxy-2-fluoro-D-glucose, FDG
About This Item
grade
pharmaceutical primary standard
API family
fludeoxyglucose
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
OCC1OC(O)C(F)C(O)C1O
InChI
1S/C6H11FO5/c7-3-5(10)4(9)2(1-8)12-6(3)11/h2-6,8-11H,1H2
InChI key
ZCXUVYAZINUVJD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
- PET Imaging in Lung Cancer: Fludeoxyglucose F18, used in PET scans, enhances the detection of occult lymph node metastases in lung cancer patients, offering significant improvements in diagnostic accuracy and patient management strategies (Guglielmo et al., 2023).
- Advanced Imaging Techniques: Research on time-of-flight (TOF) and non-TOF PET imaging explores the validation of reconstruction algorithms with NEMA phantoms, underscoring the critical role of Fludeoxyglucose in refining diagnostic approaches in clinical settings (Kumar et al., 2022).
- Fibroblast Activation Protein Inhibitor (FAPI) PET/CT Imaging: Non-oncologic incidental uptake studies using FAPI PET/CT imaging highlight Fludeoxyglucose′s versatility as a tracer that can potentially identify and characterize a variety of disease processes beyond its primary application in oncology (Hotta et al., 2023).
- Impact on HPV-related Cancer Diagnosis: Fludeoxyglucose is instrumental in imaging studies of human papilloma virus-associated oropharyngeal squamous cell carcinoma, aiding in the diagnosis and monitoring of this increasingly prevalent cancer, which is linked to significant morbidity and mortality rates (Touska et al., 2022).
Analysis Note
Other Notes
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service