16018AST
Astec® CHIROBIOTIC® T2 Chiral (5 μm) HPLC Columns
L × I.D. 10 cm × 2.1 mm, HPLC Column
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52
Recommended Products
Product Name
Astec® CHIROBIOTIC® T2 Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
material
stainless steel column
Agency
suitable for USP L63
description
HPLC column
product line
Astec®
packaging
pkg of 1 ea
manufacturer/tradename
Astec®
parameter
0-45 °C temperature
241 bar pressure (3500 psi)
technique(s)
HPLC: suitable
LC/MS: suitable
L × I.D.
10 cm × 2.1 mm
matrix
high-purity silica gel particle platform
fully porous particle
particle size
5 μm
pore size
200 Å
operating pH range
3.8-6.8
separation technique
chiral
Looking for similar products? Visit Product Comparison Guide
General description
CHIROBIOTIC® T and T2 have teicoplanin as the chiral selector. They offer unique selectivity for a number of classes of molecules, specifically underivatized α, β, γ and cyclic amino acids, N-derivatized amino acids, hydroxy-carboxylic acids, acidic compounds including carboxylic acids and phenols, small peptides, neutral aromatic analytes and cyclic aromatic and aliphatic amines. Separations normally obtained on a chiral crown ether or ligand exchange-type CSPs are also possible on the CHIROBIOTIC® T and T2, but in much simpler mobile phases, like water-alcohol. In addition, all of the known beta-blockers (amino alcohols), and dihydrocoumarins have been resolved. CHIROBIOTIC® T and T2 differ in their bonding chemistry and the pore size of the support particle, giving them different selectivity and preparative capacity.
- Bonded phase: Teicoplanin
- Operating pH range: 3.8 - 6.8
- Particle diameter: 5, 10 or 16 μm
- Pore size: 100 Å (CHIROBIOTIC® T) or 200 Å (CHIROBIOTIC® T2)
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Chromatography Liquid Chiral Separations
Xiao, T.L., et al.
Encyclopedia of Separation Science, 1-15 (2000)
Chromatographic Separations and Analysis: Macrocyclic Glycopeptide Chiral Stationary Phases
Berthod, A.
Comprehensive Chirality, 8, 227-262 (2012)
Ping Sun et al.
Journal of molecular structure, 890(1-3), 75-80 (2008-11-12)
A high performance liquid chromatographic method using macrocyclic glycopeptide chiral stationary phases (CSPs) was used to separate enantiomers of seven ruthenium(II) polypyridyl complexes. Among the five different CSPs, the Chirobiotic T2 was most effective and baseline separated all complexes. All
Chris Hamman et al.
Journal of chromatography. A, 1305, 310-319 (2013-07-31)
A rapid screening method to identify the best conditions for chiral separations is described. We analyzed a representative set of 80 racemic compounds against 25 different chiral stationary phases with three different mobile phases to identify the combination of columns
István Ilisz et al.
Journal of chromatography. A, 1269, 94-121 (2012-07-28)
Amino acids are essential for life, and have many functions in metabolism. One particularly important function is to serve as the building blocks of peptides and proteins, giving rise complex three dimensional structures through disulfide bonds or crosslinked amino acids.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service