Skip to Content
Merck
All Photos(9)

Key Documents

ZRB2975

Sigma-Aldrich

Anti-Tenascin Antibody, clone 1G4 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Cytotactin, GMEM, GP150-225, Glioma-associated-extracellular matrix antigen, Hexabrachion, JI, Myotendinous antigen, Neuronectin, TN, TN-C, Tenascin-C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
conjugate:
unconjugated
application:
IHC
WB
affinity binding assay
clone:
recombinant monoclonal
species reactivity:
human, mouse, rat
citations:
technique(s):
affinity binding assay: suitable
immunohistochemistry: suitable
western blot: suitable

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

recombinant monoclonal

description

Clone, 1G4

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 240.85 kDa
observed mol wt ~241 kDa

purified by

using Protein A

species reactivity

human, mouse, rat

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... TNC(3371)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1G4 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Tenascin. It targets an epitope within 20 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 20 amino acids from the C-terminal region of human Tenascin.

Application

Quality Control Testing

Evaluated by Western Blotting in SK-MEL-28 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Tenascin in SK-MEL-28 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Tenascin in lysates from C6 cells and Mouse brain tissue and 1:10,000 dilution in Human brain tissue.

Affinity Binding Assay: A representative lot of this antibody bound Tenascin peptide with a KD of 1.7 x 10-7KD of 1.7 x 10-7 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Tenascin in Human uterus and Mouse embryonic tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Tenascin (UniProt: P24821; also known as TN, Cytotactin, GMEM, GP 150-225, Glioma-associated-extracellular matrix antigen, Hexabrachion, JI, Myotendinous antigen, Neuronectin, Tenascin-C, TN-C) is encoded by the TNC (also known as HXB) gene (Gene ID: 3371) in human. Tenascins are a family of large multimeric extracellular matrix (ECM) proteins. In vertebrates four types of tenascins have been described: C, -R, -X and -W and each of these have a specific expression pattern. They are synthesized primarily by cells in connective tissues. However, each of the tenascins has a typical expression pattern that differs from all other tenascins. Tenascin-C is a highly conserved protein that is present in the central nervous system and is a prominent constituent of peripheral nerves. It is implicated in guidance of migrating neurons as well as axons during development, synaptic plasticity, and neuronal regeneration. It also promotes neurite outgrowth from cortical neurons grown on a monolayer of astrocytes. It inhibits cell adhesion of many cell types to fibronectin. Tenascin-C is a hexameric, multimodular ECM protein with several molecular forms that are created through alternative splicing and protein modifications. It is synthesized with a signal peptide (aa 1-22) that is subsequently cleaved off in the mature protein. It contains heptad repeats near its N-terminus that support trimerization and two trimers can join to form a hexabrachion. The heptad repeats are followed from N- to C-terminus by one or more EGF-like domains, a string of fibronectin-type III repeats and a fibrinogen-like globe. This multimodular structure enables it to interact with a high number of highly diverse ligands. Tenascin-C is highly induced by infections and inflammation and tenascin-C knockout mouse exhibit reduced inflammatory response. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Midwood, KS., et al. (2016). J. Cell Sci. 129(23); 4321-4327; Chiquet-Ehrismann, R. (2004). Int. J. Biochem. Cell Biol. 36(6); 986-990).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service