Skip to Content
Merck
All Photos(1)

Key Documents

Y4625

Sigma-Aldrich

Yeast Brewers

Debittered, BioReagent, suitable for insect cell culture

Synonym(s):

Yeast, brewers, Brewers yeast

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352205
NACRES:
NA.75

Quality Level

product line

BioReagent

form

powder

technique(s)

cell culture | insect: suitable

solubility

0.1 M sodium bicarbonate: 200 mg/mL, turbid

shipped in

ambient

storage temp.

2-8°C

SMILES string

OC(=O)CC1c2c3c(ccc2c4c1cccc4)cccc3

InChI

1S/C19H14O2/c20-18(21)11-17-15-8-4-3-7-14(15)16-10-9-12-5-1-2-6-13(12)19(16)17/h1-10,17H,11H2,(H,20,21)

InChI key

GQNBDGXKDJSVGQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eric Le Bourg et al.
Experimental gerontology, 69, 202-210 (2015-07-15)
Many experiments have shown that dietary restriction, for instance by removing live yeast or modifying the protein/carbohydrate ratio, can modulate lifespan, fecundity, resistance to severe stresses and behaviour in Drosophila melanogaster flies. The present study tested whether feeding flies with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service