Skip to Content
Merck
All Photos(3)

Key Documents

T1258

Sigma-Aldrich

Trizma® acetate

≥99.0% (titration)

Synonym(s):

TRIS acetate salt, Tris(hydroxymethyl)aminomethane acetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(CH2OH)3 · CH3COOH
CAS Number:
Molecular Weight:
181.19
Beilstein:
3702918
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

Assay

≥99.0% (titration)

form

crystalline powder

pH

6.0-7.0 (0.1 M in water, high purity)

solubility

water: 0.5 M, clear, colorless

SMILES string

CC(O)=O.NC(CO)(CO)CO

InChI

1S/C4H11NO3.C2H4O2/c5-4(1-6,2-7)3-8;1-2(3)4/h6-8H,1-3,5H2;1H3,(H,3,4)

InChI key

PIEPQKCYPFFYMG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Trizma® acetate has been used:
  • in the adenosine triphosphate (ATP) assay to neutralize the pH to 7.7 of the oocyte supernatant
  • in cell wall isolation to remove proteins and membranes from the cell walls of plants
  • as a component of DamID buffer, lysis buffer for cell sorting and lysing

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Máté Borsos et al.
Nature, 569(7758), 729-733 (2019-05-24)
In mammals, the emergence of totipotency after fertilization involves extensive rearrangements of the spatial positioning of the genome1,2. However, the contribution of spatial genome organization to the regulation of developmental programs is unclear3. Here we generate high-resolution maps of genomic
S Hashimoto et al.
Molecular reproduction and development, 57(4), 353-360 (2000-11-07)
The effects of carbohydrates on meiotic maturation and ATP content of bovine oocytes under low oxygen tension (5%) were investigated. Furthermore, the developmental competence or intracellular H(2)O(2) contents of the oocytes matured under 5% or 20% O(2) was assessed. In
Masanobu Yamamoto et al.
ChemSusChem, 13(17), 4400-4408 (2020-07-22)
Increases in coniferaldehyde content, a minor lignin residue, significantly improves the sustainable use of plant biomass for feed, pulping, and biorefinery without affecting plant growth and yields. Herein, different analytical methods are compared and validated to distinguish coniferaldehyde from other
Máté Borsos et al.
Nature, 569(7758), 729-733 (2019-05-24)
In mammals, the emergence of totipotency after fertilization involves extensive rearrangements of the spatial positioning of the genome1,2. However, the contribution of spatial genome organization to the regulation of developmental programs is unclear3. Here we generate high-resolution maps of genomic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service