SRP5193
IRS1 (1-355), GST tagged human
recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
HIRS-1
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.32
Recommended Products
recombinant
expressed in E. coli
Assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
mol wt
~65 kDa
NCBI accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... IRS1(3667)
General description
IRS1 is the substrate for the insulin tyrosine kinase receptor and is found in a variety of insulin-responsive cells and tissues. IRS1 protein has no intrinsic enzymatic activity but acts as a docking protein, via the SH2 domains, for mediating the insulin downstream signaling events. IRS1 has been shown to associate with the 14-3-3 family of proteins and this could play a role in the regulation of insulin sensitivity by interrupting the association between the insulin receptor and IRS1. IRS1 may be associated with colorectal cancer and diet and related factors may affect the risk by modifying plasma insulin levels. Thus, the inter-individual variation in insulin signaling mediated by IRS1 may play a plausible role in the development of colorectal cancer.
Physical form
Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.
Preparation Note
after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
T Ogihara et al.
The Journal of biological chemistry, 272(40), 25267-25274 (1997-10-06)
Insulin binding to its receptor induces the phosphorylation of cytosolic substrates, insulin receptor substrate (IRS)-1 and IRS-2, which associate with several Src homology-2 domain-containing proteins. To identify unique IRS-1-binding proteins, we screened a human heart cDNA library with 32P-labeled recombinant
M L Slattery et al.
Breast cancer research and treatment, 104(2), 197-209 (2006-10-20)
An insulin-related pathway to breast cancer has been hypothesized. We examine the 19 CA repeat of the IGF1 gene, the -202 C > A IGFBP3, the G972R IRS, and the G1057D IRS2 polymorphisms among 1,175 non-Hispanic white (NHW) and 576
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service