Skip to Content
Merck
All Photos(2)

Key Documents

SML3688

Sigma-Aldrich

Hypoxia probe 4

≥95% (HPLC)

Synonym(s):

5-Methoxy-1,2-dimethyl-3-(((3-oxo-9-(O-tolyl)-3H-xanthen-6-yl)oxy)methyl)-1H-indole-4,7-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H25NO6
Molecular Weight:
519.54
UNSPSC Code:
41116158
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

storage condition

protect from light

color

, Faint yellow to yellow-brown

solubility

DMSO: 2 mg/mL, clear

shipped in

wet ice

storage temp.

-10 to -25°C

SMILES string

CN1C(C(C=C2OC)=O)=C(C2=O)C(COC3=CC=C(C(C4=CC=CC=C4C)=C5C=C6)C(OC5=CC6=O)=C3)=C1C

InChI

1S/C32H25NO6/c1-17-7-5-6-8-21(17)29-22-11-9-19(34)13-26(22)39-27-14-20(10-12-23(27)29)38-16-24-18(2)33(3)31-25(35)15-28(37-4)32(36)30(24)31/h5-15H,16H2,1-4H3

InChI key

MQKQLIOGOATWPT-UHFFFAOYSA-N

Biochem/physiol Actions

Hypoxia probe 4 is a Tokyo green-based non-fluorescent hypoxia probe that undergoes cytochrome P450 reductase (PH51)-catalyzed reduction to yield the fluorescent reporter Tokyo green (ex 480 nm, em 515 nm) in conditions of 0.5% O2 and lower, allowing imaging hypoxia levels similar to those detected by EF5 without the use of antibodies. When employed together with resorufin-based hypoxia probe 2 (ex 545 nm, em 582 nm), the probe pair allows the simultaneous (two-color) imaging of different hypoxia levels in living cells.
Tokyo green-based hypoxia probe that allows fluorescent imaging of hypoxia levels in live cells similar to those detected by EF5 without the use of antibodies.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Two Color Imaging of Different Hypoxia Levels in Cancer Cells
Journal of the American Chemical Society, 145(4), 2572-2583 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service