Skip to Content
Merck
All Photos(1)

Documents

SML3507

Sigma-Aldrich

Hirsutine

≥98% (HPLC)

Synonym(s):

(αE,​2S,​3R,​12bR)​-3-​Ethyl-​1,​2,​3,​4,​6,​7,​12,​12b-​octahydro-​α-​(methoxymethylene)​-indolo[2,​3-​a]​quinolizine-​2-​acetic acid methyl ester, (E)-16,17-Didehydro-17-methoxy-17,18-seco-3-beta-yohimban-16-carboxylic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28N2O3
CAS Number:
Molecular Weight:
368.47
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

CC[C@@]1([H])CN(CCC2=C3NC4=C2C=CC=C4)[C@]3([H])C[C@]1([H])/C(C(OC)=O)=C\OC

InChI

1S/C22H28N2O3/c1-4-14-12-24-10-9-16-15-7-5-6-8-19(15)23-21(16)20(24)11-17(14)18(13-26-2)22(25)27-3/h5-8,13-14,17,20,23H,4,9-12H2,1-3H3/b18-13+/t14-,17-,20+/m0/s1

InChI key

NMLUOJBSAYAYEM-AZQGJTAVSA-N

Biochem/physiol Actions

Hirsutine is a brain penetrant alkaloid from Uncaria rhynchophylla that exhibit anti-cancer, cardioprotective, anti-hypertensive, and anti-arrhythmic activities. It appears that hirsutine induces apoptosis in lung cancer cells through opening of mitochondrial permeability transition pore (mPTP). Hirsutine exhibit potent anti-dengue virus activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Blood-Brain Barrier Permeability of Six Indole Alkaloids from Uncariae Ramulus Cum Uncis in the MDCK-pHaMDR Cell Monolayer Model.
Zhang YN, Yang YF, Xu W, Yang XW
Biomolecules, 22, 1944-1944 (2017)
Takayuki Hishiki et al.
Frontiers in microbiology, 8, 1674-1674 (2017-09-16)
Dengue virus (DENV) is transmitted to humans by Aedes mosquitoes and is a public health issue worldwide. No antiviral drugs specific for treating dengue infection are currently available. To identify novel DENV inhibitors, we analyzed a library of 95 compounds
Rong Zhang et al.
Cell death & disease, 9(6), 598-598 (2018-05-24)
Hirsutine extracted from Uncaria rhynchophylla has been shown to exhibit anti-cancer activity. However, the molecular mechanism by which hirsutine exhibits anti-lung cancer activity remains unclear. In the present study, we showed that hirsutine induces apoptosis in human lung cancer cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service