Skip to Content
Merck
All Photos(1)

Key Documents

SML2842

Sigma-Aldrich

BAY 1125976

≥98% (HPLC)

Synonym(s):

2-[4-(1-Aminocyclobutyl)phenyl]-3-phenylimidazo[1,2-b]pyridazine-6-carboxamide, BAY-1125976, BAY1125976

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H21N5O
CAS Number:
Molecular Weight:
383.45
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[n]21nc(ccc2nc(c1c5ccccc5)c3ccc(cc3)C4(CCC4)N)C(=O)N

InChI

1S/C23H21N5O/c24-22(29)18-11-12-19-26-20(21(28(19)27-18)16-5-2-1-3-6-16)15-7-9-17(10-8-15)23(25)13-4-14-23/h1-3,5-12H,4,13-14,25H2,(H2,24,29)

InChI key

JBGYKRAZYDNCNV-UHFFFAOYSA-N

Biochem/physiol Actions

BAY 1125976 is an orally active, potent and highly selective AKT1/2 allosteric inhibitor (IC50 = 5.2/18 nM with 10 μM ATP, IC50 = 44/36 nM with 2 mM ATP; AKT3 IC50 = 427 nM with 10 μM ATP) that targets a pocket formed by the kinase & PH domain. BAY 1125976 reduces basal AKT T308/S473 phosphorylation (IC50 = 0.9/1.1; KPL-4 cells) and exhibits anti-cancer efficacy in cultures (IC50 = 0.02-10 μM, 23 human cancer lines) and in xenograft models in vivo (25, 50 mg/kg/day or 5, 15, 25 mg/kg bid. po.; mice), including breast cancers (KPL4 PIK3CAH1074R mutant, MCF7, HBCx-2), AKTE17K-driven prostate cancer LAPC-4, and anal cancer AXF 984.
Orally active, potent and highly selective AKT1/2 allosteric inhibitor with anti-cancer efficacy in cultures and in murine xenograft models in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oliver Politz et al.
International journal of cancer, 140(2), 449-459 (2016-10-22)
The PI3K-AKT-mTOR signaling cascade is activated in the majority of human cancers, and its activation also plays a key role in resistance to chemo and targeted therapeutics. In particular, in both breast and prostate cancer, increased AKT pathway activity is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service