Skip to Content
Merck
All Photos(1)

Key Documents

SML1883

Sigma-Aldrich

AR231453

≥98% (HPLC)

Synonym(s):

AR231453, (2-Fluoro-4-methanesulfonylphenyl)-(6-[4-(3-isopropyl-[1,2,4]oxadiazol-5-yl)-piperidin-1-yl]-5-nitropyrimidin-4-yl)amine, AR-231,453, N-(2-Fluoro-4-(methylsulfonyl)phenyl)-6-(4-(3-isopropyl-1,2,4-oxadiazol-5-yl)piperidin-1-yl)-5-nitropyrimidin-4-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H24FN7O5S
CAS Number:
Molecular Weight:
505.52
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

CC(C)C1=NOC(C(CC2)CCN2C(N=CN=C3NC4=CC=C(C=C4F)S(=O)(C)=O)=C3[N+]([O-])=O)=N1

InChI

1S/C21H24FN7O5S/c1-12(2)18-26-21(34-27-18)13-6-8-28(9-7-13)20-17(29(30)31)19(23-11-24-20)25-16-5-4-14(10-15(16)22)35(3,32)33/h4-5,10-13H,6-9H2,1-3H3,(H,23,24,25)

InChI key

DGBKNTVAKIFYNU-UHFFFAOYSA-N

Biochem/physiol Actions

AR231453 is a 5-nitropyrimidine derivative that acts as a highly potent and selective GPR119 agonist (EC50/GPR119 species = 0.4 nM/monkey, 1.6 nM/dog, 4.7 nM/hamster, 3.5 nM/human, 12 nM/mouse, 4 nM/rat in cell-based assays), exhibiting no off-target activity toward 216 receptors, enzymes, and orphan GPCRs. AR231453 improves glucose tolerance in mice (3 mg/kg i.p. or 20 mg/kg, p.o.) and rats (3 mg/kg i.p.), while no in vivo efficacy is observed among GPR119-deficient mice.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhi-Liang Chu et al.
Endocrinology, 148(6), 2601-2609 (2007-02-10)
Pancreatic beta-cell dysfunction is a hallmark event in the pathogenesis of type 2 diabetes. Injectable peptide agonists of the glucagon-like peptide 1 (GLP-1) receptor have shown significant promise as antidiabetic agents by virtue of their ability to amplify glucose-dependent insulin
Christoffer Norn et al.
Structure (London, England : 1993), 23(12), 2377-2386 (2015-11-04)
Recent benchmark studies have demonstrated the difficulties in obtaining accurate predictions of ligand binding conformations to comparative models of G-protein-coupled receptors. We have developed a data-driven optimization protocol, which integrates mutational data and structural information from multiple X-ray receptor structures
Graeme Semple et al.
Journal of medicinal chemistry, 51(17), 5172-5175 (2008-08-14)
GPR119 is a rhodopsin-like GPCR expressed in pancreatic beta-cells and incretin releasing cells in the GI tract. As with incretins, GPR119 increases cAMP levels in these cell types, thus making it a highly attractive potential target for the treatment of
Deborah A Goldspink et al.
Cell reports, 31(13), 107833-107833 (2020-07-02)
Glucagon-like peptide-1 (GLP-1) from intestinal L-cells stimulates insulin secretion and reduces appetite after food ingestion, and it is the basis for drugs against type-2 diabetes and obesity. Drugs targeting L- and other enteroendocrine cells are under development, with the aim

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service