Skip to Content
Merck
All Photos(1)

Key Documents

SML0702

Sigma-Aldrich

MRT67307 hydrochloride

≥98% (HPLC)

Synonym(s):

N-[3-[[5-Cyclopropyl-2-[[3-(4-morpholinylmethyl)phenyl]amino]-4-pyrimidinyl]amino]propyl]-cyclobutanecarboxamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H36N6O2 · xHCl
CAS Number:
Molecular Weight:
464.60 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to light brown

solubility

H2O: 15 mg/mL, clear

storage temp.

2-8°C

SMILES string

N5(CCOCC5)Cc1cc(ccc1)Nc2nc(c(cn2)C4CC4)NCCCNC(=O)C3CCC3

InChI

1S/C26H36N6O2/c33-25(21-5-2-6-21)28-11-3-10-27-24-23(20-8-9-20)17-29-26(31-24)30-22-7-1-4-19(16-22)18-32-12-14-34-15-13-32/h1,4,7,16-17,20-21H,2-3,5-6,8-15,18H2,(H,28,33)(H2,27,29,30,31)

InChI key

UKBGBACORPRCGG-UHFFFAOYSA-N

Application

MRT67307 hydrochloride has been used to study its effect on LPS (lipopolysaccharide) -induced lysosome tubulation.

Biochem/physiol Actions

MRT67307 is a dual inhibitor of the IKKe and TBK-1.
MRT67307 is a dual inhibitor of the IKKe and TBK-1. IKKe and TBK-1 mediate the phosphorylation of interferon regulatory factor 3 (IRF3).
MRT67307 is an amino-pyrimidine derivative and has an IC50 value of 19. It is known to induce TLR (toll like receptor) mediated anti inflammatory cytokine generation. Also, MRT67307 prevents the secretion of cytokines associated with proinflammation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

mTOR controls lysosome tubulation and antigen presentation in macrophages and dendritic cells.
Saric A, et al.
Molecular Biology of the Cell, 27(2), 321-333 (2016)
Daniel P.
The Inhibitor Index: A Desk Reference on Enzyme Inhibitors, Receptor Antagonists, Drugs, Toxins, Poisons, Biologics, and Therapeutic Leads (2017)
Zachary P Guinn et al.
Immunobiology, 224(4), 565-574 (2019-05-11)
IFN-γ produced during viral infections activates the IFN-γ receptor (IFNGR) complex for STAT1 transcriptional activity leading to expression of Interferon Regulatory Factors (IRF). Simultaneous activation of TBK/IKKε via TLR3 during viral infections activates the transcription factor IRF3. Together these transcription factors
Therapeutic potential of targeting TBK1 in autoimmune diseases and interferonopathies.
Hasan M and Nan Y
Pharmacological Research, 111, 336-342 (2016)
Noémie Pied et al.
PLoS pathogens, 18(7), e1010736-e1010736 (2022-07-21)
Intracellular pathogens cause membrane distortion and damage as they enter host cells. Cells perceive these membrane alterations as danger signals and respond by activating autophagy. This response has primarily been studied during bacterial invasion, and only rarely in viral infections.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service