SCJ4600026
CF™ 568 maleimide
suitable for fluorescence, ≥90% (TLC)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
Recommended Products
Quality Level
Assay
≥90% (TLC)
form
solid
mol wt
~836 g/mol
solubility
DMF: soluble
DMSO: soluble
H2O: soluble
ε (extinction coefficient)
100000 at 562 nm
absorbance ratio
A280/562 nm 0.08
fluorescence
λex 488,532 or 561 nm (Flow cytometry laser line)
λex 562 nm; λem 583 nm
λex 568 nm (Microscopy laser line)
suitability
suitable for fluorescence
shipped in
wet ice
storage temp.
−20°C
Related Categories
General description
CF™568 is a red fluorescent dye with an excitation spectrum optimally matching the 568 nm line of the Ar-Kr mixed-gas laser. Antibody conjugates of CF™568 are much brighter than those of Alexa Fluor™ 568. In addition, the photostability of CF™568 is superior to that of Alexa Fluor™ 568, making CF™568 a much better choice for demanding applications such as confocal microscopy and single molecule imaging.
Features and Benefits
1. Yields much brighter antibody conjugates than Alexa Fluor™568. 2. Extremely photostable. 3. Highly water-soluble.
Preparation Note
Protect from light
Reconstitution
Dissolve in DMSO
Legal Information
This product is distributed by Sigma-Aldrich Co. under the authorization of Biotium, Inc. This product is covered by one or more US patents and corresponding patent claims outside the US patents or pending applications owned or licensed by Biotium, Inc. including without limitation: 12/334,387; 12/607,915; 12/699,778; 12/850,578; 61/454,484. In consideration of the purchase price paid by the buyer, the buyer is hereby granted a limited, non-exclusive, non-transferable license to use only the purchased amount of the product solely for the buyer′s own internal research in a manner consistent with the accompanying product literature. Except as expressly granted herein, the sale of this product does not grant to or convey upon the buyer any license, expressly, by implication or estoppel, under any patent right or other intellectual property right of Biotium, Inc. Buyer shall not resell or transfer this product to any third party, or use the product for any commercial purposes, including without limitation, any diagnostic, therapeutic or prophylactic uses. This product is for research use only. Any other uses, including diagnostic uses, require a separate license from Biotium, Inc. For information on purchasing a license to use this product for purposes other than research, contact Biotium, Inc., 3159 Corporate Place, Hayward, CA 94545, Tel: (510) 265-1027. Fax: (510) 265-1352. Email: [email protected].
ALEXA FLUOR is a trademark of Life Technologies
CF is a trademark of Biotium, Inc.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service