Skip to Content
Merck
All Photos(2)

Key Documents

SAB4502034

Sigma-Aldrich

Anti-TRK B antibody produced in rabbit

affinity isolated antibody

Synonym(s):

BDNF/NT-3 growth factors receptor precursor, GP145-TrkB, GP145-TrkB/GP95-TrkB, NTRK2, Trk-B

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 91 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NTRK2(4915)

General description

Anti-TRK B Antibody detects endogenous levels of total TRK B protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human Trk B.

Immunogen Range: 481-530

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kaige Ma et al.
International journal of biological sciences, 19(2), 675-690 (2023-01-13)
Pain is the major reason that patients suffering from osteoarthritis (OA) seek medical care. We found that vascular endothelial growth factors (VEGFs) mediate signaling in OA pain pathways. To determine the specific contributions of VEGFs and their receptors (VEGFRs) to
Myeongjeong Choo et al.
Nature communications, 8(1), 195-195 (2017-08-05)
Elimination of early-formed redundant synapses during postnatal development is essential for functional neural circuit formation. Purkinje cells (PCs) in the neonatal cerebellum are innervated by multiple climbing fibers (CFs). A single CF is strengthened whereas the other CFs are eliminated
Yan Liu et al.
Neuropharmacology, 144, 337-344 (2018-11-09)
NSI-189 Phosphate, (4-benzylpiperazin-1-yl)-[2-(3-methyl-butylamino)pyridin-3-yl] methanone is a new chemical entity under development for the treatment of MDD, based upon preclinical data demonstrating stimulation of neurogenesis of human hippocampus-derived neural stem cells in vitro and in mouse hippocampus in vivo. Previous studies
Arjun Singh et al.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society, 34(4), e14099-e14099 (2021-02-25)
Data on the neuromodulatory effects of brain-derived neurotrophic factor (BDNF) in the gastrointestinal tract were recently reported, but there are still no data on the presence, distribution, and release of BDNF in the gastrointestinal tract, including the internal anal sphincter

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service