Skip to Content
Merck
All Photos(3)

Key Documents

SAB4500078

Sigma-Aldrich

Anti-p15 INK antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CDKN2B, CDN2B, Cyclin-dependent kinase 4 inhibitor B, MTS2, Multiple tumor suppressor 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 14 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDKN2B(1030)

General description

Anti-p15 INK Antibody detects endogenous levels of total p15 INK protein.
Cyclin-dependent kinase inhibitor 2B (CDKN2B) gene codes for tumor-suppressor p15INK4B protein. p15INK4B is a member of the INK4 family of protein kinase inhibitors. In bone marrow cells, p15INK4B expression is mainly seen at a high level in maturing monocytes/macrophages and lymphocytes. It is located in the nucleus and cytoplasm. The CDKN2B gene is mapped to human chromosome 9p21.3. Anti-p15 INK Antibody detects endogenous levels of total p15 INK protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human p15 INK.

Immunogen Range: 89-138

Application

Anti-p15 INK antibody produced in rabbit has been used in western blot (1:500).

Biochem/physiol Actions

Cyclin-dependent kinase inhibitor 2B (CDKN2B) plays a key role in the cell cycle and senescence. It participates in the differentiation of the hematopoietic cell. CDKN2B gene is involved in the pathogenesis of T-acute lymphoblastic leukemia (T-ALL). Homozygous deletion of the CDKN2B gene or p15INK4B locus on chromosome results in glioblastoma multiforme (GBM). Silencing of p15INK4B is associated with myelodysplastic syndromes, leukemia, and myeloproliferative diseases.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CDKN2B (cyclin-dependent kinase inhibitor 2B (p15, inhibits CDK4))
Fares J, et al.
Atlas of Genetics and Cytogenetics in Oncology and Haematology (2011)
Woori Jang et al.
Experimental & molecular medicine, 51(1), 1-15 (2019-01-13)
We identified principal genetic alterations in 97.1% (99/102) of patients with T-acute lymphoblastic leukemia (T-ALL) using integrative genetic analyses, including massive parallel sequencing and multiplex ligation-dependent probe amplification (MLPA). A total of 133 mutations were identified in the following genes
Soon Sang Park et al.
Heliyon, 9(2), e13170-e13170 (2023-02-15)
Senescent tumor cells are nonproliferating tumor cells which are closely related to cancer progression by secreting senescence-related molecules, called senescence-associated secreting phenotypes. Therefore, the presence of senescent tumor cells is considered a prognostic factor in various cancer types. Although senescence-associated
Martin S Minns et al.
The American journal of pathology, 186(2), 285-296 (2015-12-20)
The process of wound healing involves a complex network of signaling pathways working to promote rapid cell migration and wound closure. Activation of purinergic receptors by secreted nucleotides plays a major role in calcium mobilization and the subsequent calcium-dependent signaling
Anthony Lagnado et al.
The EMBO journal, 40(9), e106048-e106048 (2021-03-26)
Cellular senescence is characterized by an irreversible cell cycle arrest as well as a pro-inflammatory phenotype, thought to contribute to aging and age-related diseases. Neutrophils have essential roles in inflammatory responses; however, in certain contexts their abundance is associated with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service