Skip to Content
Merck
All Photos(1)

Key Documents

SAB4300656

Sigma-Aldrich

Anti-phospho-VEGFR2 (pTyr1059) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CD309, Anti-FLK1, Anti-KDR, Anti-VEGFR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~230 kDa

species reactivity

mouse, human

concentration

1 mg/mL

technique(s)

western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(P-D-Yp-V-R)

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr1059)

Gene Information

human ... VEGFR2(3791)

General description

The vascular endothelial growth factor receptor-2 (VEGFR-2) is also called as kinase insert domain containing receptor (KDR). There are seven immunoglobulin-like (Ig-like) domains at the extracellular region (EC) of VEGFR-2. This gene is mapped to human chromosome 4q12.

Immunogen

Peptide sequence around phosphorylation site of tyrosine 1059 (P-D-Yp-V-R), according to the protein NP_002244.1

Biochem/physiol Actions

The immunoglobulin-like (Ig-like) domains of the vascular endothelial growth factor receptor-2 (VEGFR-2) are essential for specific ligand binding and receptor dimerization. It helps to activate multiple signaling cascades. VEGF participates in angiogenesis. VEGFR2 is involved in progression of several cancers like gliomas, malignant peripheral nerve sheath tumors, triple-negative breast cancer (TNBC) and gastrointestinal stromal tumors (GISTs).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Receptor for VEGF or VEGFC. Has a tyrosine-protein kinase activity. The VEGF-kinase ligand/receptor signaling system plays a key role in vascular development and regulation of vascular permeability. In case of HIV-1 infection, the interaction with extracellular viral Tat protein seems to enhance angiogenesis in Kaposi′s sarcoma lesions

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Immunoglobulin-like domain 4-mediated ligand-independent dimerization triggers VEGFR-2 activation in HUVECs and VEGFR2-positive breast cancer cells.
Zhang S, et al.
Breast Cancer Research and Treatment, 163(3), 423-434 (2017)
Vascular endothelial growth factor receptor-2 polymorphisms have protective effect against the development of tendinopathy in volleyball athletes.
Salles JI, et al.
PLoS ONE, 11(12), e0167717-e0167717 (2016)
The three receptor tyrosine kinases c-KIT, VEGFR2 and PDGFRα, closely spaced at 4q12, show increased protein expression in triple-negative breast cancer.
Jansson S, et al.
PLoS ONE, 9(7), e102176-e102176 (2014)
Characterization of the amplicon on chromosomal segment 4q12 in glioblastoma multiforme.
Holtkamp N, et al.
Neuro-Oncology, 9(3), 291-297 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service