Skip to Content
Merck
All Photos(1)

Key Documents

SAB4200743

Sigma-Aldrich

Anti-Human IgG (Fc specific)-Peroxidase antibody, Mouse monoclonal

clone HP-6017, purified from hybridoma cell culture

Synonym(s):

Anti-Immunoglubulin G (Fc specific)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

Quality Level

conjugate

peroxidase conjugate

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

HP-6017, monoclonal

form

lyophilized powder

species reactivity

human

concentration

~2 mg/mL

technique(s)

ELISA: 1:80,000-1:160,000 using 5 μg/mL Human IgG myeloma for coating

isotype

IgG2a

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Anti-Human IgG (Fc specific)-Peroxidase antibody, Mouse monoclonal (mouse IgG2a isotype) is derived from the HP-6017 hybridoma, produced by the fusion of mouse myeloma cells and splenocytes from a mouse immunized with purified human IgG myeloma proteins covalently coupled to poly aminostyrene (PAS) microbeads. IgGs (Immunoglobulin G) are the most common Immunoglobulin′s isotype in blood, lymph, cerebrospinal and peritoneal fluids and serve as key players in the humoral immune response. IgGs include four subclasses (IgG1, IgG2, IgG3, and IgG4), consist of a variable Fab fragment (which includes the antibody recognition site) and a conserved Fc fragment.

Immunogen

Human IgG myeloma proteins

Application

Anti-Human IgG (Fc specific)-Peroxidase antibody has been used in enzyme linked immunosorbent assay (ELISA).

Biochem/physiol Actions

IgG (Immunoglobulin G) deficiencies are often associated with various diseases. The Fc fragment has various important functions (e.g complement fixation, site for rheumatoid factor attachment and protein A binding) which indicate on the importance of immunoreagents specific for the Fc fragment of Human IgG.

Physical form

Supplied as a lyophilized powder.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Jefferis et al.
Immunology letters, 10(3-4), 223-252 (1985-01-01)
Seventy-four monoclonal antibodies (McAb) of putative specificity for human IgG (11), the IgG sub-classes (59) or Gm allotypes (4) have been evaluated for reactivity and specificity in eight laboratories employing different assay techniques or protocols. For the IgG, IgG3, IgG4
Gestur Vidarsson et al.
Frontiers in immunology, 5, 520-520 (2014-11-05)
Of the five immunoglobulin isotypes, immunoglobulin G (IgG) is most abundant in human serum. The four subclasses, IgG1, IgG2, IgG3, and IgG4, which are highly conserved, differ in their constant region, particularly in their hinges and upper CH2 domains. These

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service