Skip to Content
Merck
All Photos(1)

Key Documents

SAB2500976

Sigma-Aldrich

Anti-SOD1 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ALS, Anti-ALS1, Anti-Amyotrophic lateral sclerosis 1, soluble, Anti-HGNC:11179, Anti-Superoxide dismutase 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, human, canine, mouse

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SOD1(6647)

Immunogen

Peptide with sequence C-SRKHGGPKDEERH from the internal region of the protein sequence according to NP_000445.1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

So Eun Kim et al.
Molecular medicine reports, 25(1) (2021-11-20)
Although multi‑organ dysfunction is associated with the survival rate following cardiac arrest (CA), the majority of studies to date have focused on hearts and brains, and few studies have considered renal failure. The objective of the present study, therefore, was
Dongtao Wang et al.
Skeletal muscle, 8(1), 18-18 (2018-06-02)
A low-protein diet supplemented with ketoacids (LPD + KA) maintains the nutritional status of patients with chronic kidney disease (CKD). Oxidative damage and mitochondrial dysfunction associated with the upregulation of p66SHC and FoxO3a have been shown to contribute to muscle atrophy. This
Erica de Sousa et al.
Cell death & disease, 13(4), 393-393 (2022-04-23)
The voltage-dependent anion channel 1 (VDAC1) was first described as a mitochondrial porin that mediates the flux of metabolites and ions, thereby integrating both cell survival and death signals. In the nervous system, the functional roles of VDAC1 remain poorly
Maki Yoshikawa et al.
Nutrients, 13(4) (2021-04-04)
Fucoxanthin (Fx) has preventive effect against muscle atrophy and myotube loss in vitro, but it has not yet been examined in vivo. Therefore, we aimed to investigate the effect of Fx on dexamethasone (Dex)-induced muscle atrophy and fat mass in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service