Skip to Content
Merck
All Photos(2)

Key Documents

SAB1411073

Sigma-Aldrich

Anti-TDP1 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ11090, MGC104252

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 68.4 kDa

species reactivity

human

technique(s)

immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TDP1(55775)

General description

The protein encoded by this gene is involved in repairing stalled topoisomerase I-DNA complexes by catalyzing the hydrolysis of the phosphodiester bond between the tyrosine residue of topoisomerase I and the 3-prime phosphate of DNA. This protein may also remove glycolate from single-stranded DNA containing 3-prime phosphoglycolate, suggesting a role in repair of free-radical mediated DNA double-strand breaks. This gene is a member of the phospholipase D family and contains two PLD phosphodiesterase domains. Mutations in this gene are associated with the disease spinocerebellar ataxia with axonal neuropathy (SCAN1). While several transcript variants may exist for this gene, the full-length natures of only two have been described to date. These two represent the major variants of this gene and encode the same isoform. (provided by RefSeq)

Immunogen

TDP1 (NP_001008744.1, 1 a.a. ~ 608 a.a) full-length human protein.

Sequence
MSQEGDYGRWTISSSDESEEEKPKPDKPSTSSLLCARQGAANEPRYTCSEAQKAAHKRKISPVKFSNTDSVLPPKRQKSGSQEDLGWCLSSSDDELQPEMPQKQAEKVVIKKEKDISAPNDGTAQRTENHGAPACHRLKEEEDEYETSGEGQDIWDMLDKGNPFQFYLTRVSGVKPKYNSGALHIKDILSPLFGTLVSSAQFNYCFDVDWLVKQYPPEFRKKPILLVHGDKREAKAHLHAQAKPYENISLCQAKLDIAFGTHHTKMMLLLYEEGLRVVIHTSNLIHADWHQKTQGIWLSPLYPRIADGTHKSGESPTHFKADLISYLMAYNAPSLKEWIDVIHKHDLSETNVYLIGSTPGRFQGSQKDNWGHFRLKKLLKDHASSMPNAESWPVVGQFSSVGSLGADESKWLCSEFKESMLTLGKESKTPGKSSVPLYLIYPSVENVRTSLEGYPAGGSLPYSIQTAEKQNWLHSYFHKWSAETSGRSNAMPHIKTYMRPSPDFSKIAWFLVTSANLSKAAWGALEKNGTQLMIRSYELGVLFLPSAFGLDSFKVKQKFFAGSQEPMATFPVPYDLPPELYGSKDRPWIWNIPYVKAPDTHGNMWVPS

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pooja Gupta et al.
Aging cell, 21(6), e13625-e13625 (2022-05-19)
Mutation in Werner (WRN) RECQL helicase is associated with premature aging syndrome (Werner syndrome, WS) and predisposition to multiple cancers. In patients with solid cancers, deficiency of the WRN RECQL helicase is paradoxically associated with enhanced overall survival in response

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service