Skip to Content
Merck
All Photos(4)

Key Documents

SAB1405708

Sigma-Aldrich

Anti-DDX5 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

DKFZp434E109, DKFZp686J01190, G17P1, HLR1, HUMP68, p68

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~67.54 kDa

species reactivity

mouse, human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DDX5(1655)

General description

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure, such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which is a RNA-dependent ATPase, and also a proliferation-associated nuclear antigen, specifically reacting with the simian virus 40 tumor antigen. This gene consists of 13 exons, and alternatively spliced transcripts containing several intron sequences have been detected, but no isoforms encoded by these transcripts have been identified. (provided by RefSeq)

Immunogen

DDX5 (AAH16027, 1 a.a. ~ 614 a.a) full-length human protein.

Sequence
MSGYSSDRDRGRDRGFGAPRFGGSRAGPLSGKKFGNPGEKLVKKKWNLDELPKFEKNFYQEHPDLARRTAQEVETYRRSKEITVRGHNCPKPVLNFYEANFPANVMDVIARQNFTEPTAIQAQGWPVALSGLDMVGVAQTGSGKTLSYLLPAIVHINHQPFLERGDGPICLVLAPTRELAQQVQQVAAEYCRACRLKSTCIYGGAPKGPQIRDLERGVEICIATPGRLIDFLECGKTNLRRTTYLVLDEADRMLDMGFEPQIRKIVDQIRPDRQTLMWSATWPKEVRQLAEDFLKDYIHINIGALELSANHNILQIVDVCHDVEKDEKLIRLMEEIMSEKENKTIVFVETKRRCDELTRKMRRDGWPAMGIHGDKSQQERDWVLNEFKHGKAPILIATDVASRGLDVEDVKFVINYDYPNSSEDYIHRIGRTARSTKTGTAYTFFTPNNIKQVSDLISVLREANQAINPKLLQLVEDRGSGRSRGRGGMKDDRRDRYSAGKRGGFNTFRDRENYDRGYSSLLKRDFGAKTQNGVYSAANYTNGSFGSNFVSAGIQTSFRTGNPTGTYQNGYDSTQQYGSNVPNMHNGMNQQAYAYPATAAAPMIGYPMPTGYSQ

Application

Anti-DDX5 antibody produced in mouse is suitable for indirect immunofluorescence and indirect ELISA.

Biochem/physiol Actions

DDX5 (DEAD, Asp-Glu-Ala-Asp, box helicase 5) is associated with the regulation of pre-mRNA splicing mechanism. It acts as a transcriptional coactivator to control alternative splicing during the formation of spliceosome. DDX5 provides stability to the RNA secondary structure during several activities such as in the translation initiation, nuclear and mitochondrial splicing. It facilitates the unwinding as well as detachment of protein part from RNA. The protein also participates in the estrogen- and androgen-signaling pathway.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samaan Samaan et al.
Nucleic acids research, 42(4), 2197-2207 (2013-11-28)
Estrogen and androgen receptors (ER and AR) play key roles in breast and prostate cancers, respectively, where they regulate the transcription of large arrays of genes. The activities of ER and AR are controlled by large networks of protein kinases
Frances V Fuller-Pace
Biochimica et biophysica acta, 1829(8), 756-763 (2013-03-26)
Members of the DEAD box family of RNA helicases, which are characterised by the presence of twelve conserved motifs (including the signature D-E-A-D motif) within a structurally conserved 'helicase' core, are involved in all aspects of RNA metabolism. Apart from
Ulrike Rappe et al.
The Journal of biological chemistry, 289(18), 12421-12434 (2014-03-20)
The armadillo repeat protein ARVCF is a component of adherens junctions. Similar to related proteins, such as p120-catenin and β-catenin, with known signaling functions, localization studies indicate a cytoplasmic and a nuclear pool of ARVCF. We find that ARVCF interacts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service