Skip to Content
Merck
All Photos(1)

Key Documents

SAB1300669

Sigma-Aldrich

Anti-SULT2B1a/b (N-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-Alcohol sulfotransferase, Anti-HSST2, Anti-Hydroxysteroid sulfotransferase SULT2B1b, Anti-Sulfotransferase family, cytosolic, 2B, member 1, isoform b

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SULT2B1(6820)

Related Categories

General description

Cytosolic sulfotransferases (STs or SULTs) encodes hydroxysteroid sulfotransferase (SULT2B1), the isoforms of which are SULT2B1a and SULT2B1b. By PCR with human placental and prostate cDNA as template, they isolated 2 alternatively spliced cDNAs, identical throughout most of their sequences, but having different 5-prime ends. The shorter cDNA, SULT2B1a, encodes a protein of 350 amino acids; the longer cDNA, SULT2B1b, encodes a 365-amino acid protein. Genomic PCR analysis revealed that the gene encoding both cDNAs, SULT2B1, contains 7 exons, with 2 alternative first exons being used to generate SULT2B1a and SULT2B1b. Northern blot analysis revealed that the SULT2B1 gene is expressed as a 1.4-kb transcript predominantly in prostate, placenta, and trachea. In human chromosome, the gene SULT2B1a/b is localized on 19q13.3.

Immunogen

SULT2B1A/B (NP_004596, 1-30)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the N-terminal region of human SULT2B1a/b.

Biochem/physiol Actions

Cytosolic sulfotransferases (STs or SULTs) catalyse the sulfate conjugation of many drugs, xenobiotic compounds, hormones, and neurotransmitters. SULT2B1 is involved in the synthesis and metabolism of steroids, which is essential for skin development and skin barrier function. Mutations in SULT2b1a/b gene leads to increased keratinocyte proliferation and altered cholesterol metabolism, resulting in autosomal-recessive congenital ichthyosis (ARCI). SULT2B1 catalyses the sulfation of an androgen precursor, dehydroepiandrosterone (DHEA) and hence polymorphism within this gene may lead to prostate tumor.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cytosolic sulfotransferases
Nimmagadda D, et al.
Indian Journal of Experimental Biology, 44, 171-182 (2006)
Xenobiotic-metabolizing enzymes in the skin of rat, mouse, pig, guinea pig, man, and in human skin models
Oesch F, et al.
Archives of Toxicology, 88(12), 2135-2190 (2014)
Mutations in SULT2B1 cause autosomal-recessive congenital ichthyosis in humans
Heinz L, et al.
American Journal of Human Genetics, 100(6), 926-939 (2017)
Human hydroxysteroid sulfotransferase SULT2B1: two enzymes encoded by a single chromosome 19 gene
Her C, et al.
Genomics, 53(3), 284-295 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service