Skip to Content
Merck
All Photos(1)

Documents

P1185

Sigma-Aldrich

1,2-Dipalmitoyl-sn-glycero-3-phospho-L-serine sodium salt

≥99% (TLC)

Synonym(s):

L-α-Phosphatidyl-L-serine, dipalmitoyl sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C38H73NO10PNa
CAS Number:
Molecular Weight:
757.95
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥99% (TLC)

form

powder

functional group

phospholipid

lipid type

phosphoglycerides

storage temp.

−20°C

SMILES string

[Na+].CCCCCCCCCCCCCCCC(=O)OC[C@H](COP(O)(=O)OC[C@H](N)C([O-])=O)OC(=O)CCCCCCCCCCCCCCC

InChI

1S/C38H74NO10P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-36(40)46-31-34(32-47-50(44,45)48-33-35(39)38(42)43)49-37(41)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h34-35H,3-33,39H2,1-2H3,(H,42,43)(H,44,45);/q;+1/p-1/t34-,35+;/m1./s1

InChI key

GTLXLANTBWYXGW-CEGNZRHUSA-M

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Mitra et al.
European biophysics journal : EBJ, 48(2), 119-129 (2018-11-27)
The large number of potential applications of ionic liquids (ILs) requires an understanding of their environmental impacts including their adverse effects on microorganisms living in soil and water. The molecular mechanism of toxic activities of these liquids is yet to
Éder Dos Santos Souza et al.
Analytical and bioanalytical chemistry, 412(6), 1431-1439 (2020-01-09)
Triatominae are hematophagous insects involved in the transmission of Chagas disease. Among the 19 genera of the subfamily, those with the highest epidemiological importance regarding the dissemination of Trypanosoma cruzi are Panstrongylus, Rhodnius, and Triatoma. Of these three genera, Rhodnius
Cosima D Calvano et al.
Journal of mass spectrometry : JMS, 49(9), 831-839 (2014-09-18)
In foodstuffs, one of the main factors inducing modifications in phospholipids (PLs) structure is the heat treatment. Among PLs, only phosphatidylethanolamines and phosphatidylserines, due to their free amino group, can be involved in Maillard reaction and can form adducts with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service