Skip to Content
Merck
All Photos(1)

Key Documents

P0036

Sigma-Aldrich

PP121

≥98% (HPLC)

Synonym(s):

1-cyclopentyl-3-(1H-pyrrolo[2,3-b]pyridin-5-yl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17N7
CAS Number:
Molecular Weight:
319.36
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

off-white

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

Nc1ncnc2n(nc(-c3cnc4[nH]ccc4c3)c12)C5CCCC5

InChI

1S/C17H17N7/c18-15-13-14(11-7-10-5-6-19-16(10)20-8-11)23-24(12-3-1-2-4-12)17(13)22-9-21-15/h5-9,12H,1-4H2,(H,19,20)(H2,18,21,22)

InChI key

NVRXTLZYXZNATH-UHFFFAOYSA-N

Biochem/physiol Actions

PP121 blocks the vascular endothelial growth factor (VEGF) receptor and its related signaling events. It also inhibits tumor cell proliferation by targeting oncogenic tyrosine kinase and phosphoinositide 3-kinases (PI3Ks).
PP121 is a dual inhibitor of tyrosine and phosphoinositide kinases.
PP121 is a dual inhibitor of tyrosine and phosphoinositide kinases. PP121 is the first inhibitor active on both Tyrosine kinases and the phosphatidylinositol-3-OH kinase (PI(3)K) family. The inhibitor is not effecting other serine-threonine protein kinases.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Phosphoinositide Kinases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beth Apsel et al.
Nature chemical biology, 4(11), 691-699 (2008-10-14)
The clinical success of multitargeted kinase inhibitors has stimulated efforts to identify promiscuous drugs with optimal selectivity profiles. It remains unclear to what extent such drugs can be rationally designed, particularly for combinations of targets that are structurally divergent. Here
Zhihang Zhou et al.
Experimental cell research, 388(2), 111857-111857 (2020-01-24)
Bone resorption, caused by osteoclasts (OCs), is important to bone homeostasis. The abnormalities of bone resorption may induce a series of diseases, including osteoarthritis, osteoporosis and aseptic peri-implant loosening. The latest research developed,a novel tyrosine and phosphoinositide kinase dual inhibitor

Articles

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service