Skip to Content
Merck
All Photos(1)

Key Documents

N1288

Sigma-Aldrich

NADH Oxidase from Bacillus licheniformis

lyophilized powder

Synonym(s):

NOX

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

Bacillus licheniformis

Quality Level

form

lyophilized powder

specific activity

≥35 units/mg protein

packaging

vial of ≥15 units

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

NADH Oxidase is a surface enzyme with increased oxidative activity in polymorphonuclear leukocytes during phagocytosis.

Application

NADH Oxidase from Bacillus licheniformis has been used in a study to assess nitrogen assimilation by Bacillus licheniformis growing in chemostat cultures. It has also been used in a study to investigate the role of glutamate dehydrogenase in ammonia assimilation in Bacillus macerans.

Biochem/physiol Actions

NADH Oxidase from Bacillus licheniformis was shown to display hydrogen peroxide-forming activity.

Unit Definition

One unit will oxidize 1.0 μmole NADH per minute at pH 7.0 at 30 °C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Kanamori et al.
Journal of bacteriology, 169(10), 4692-4695 (1987-10-01)
Pathways of ammonia assimilation into glutamic acid in Bacillus macerans were investigated by measurements of the specific activities of glutamate dehydrogenase (GDH), glutamine synthetase, and glutamate synthase. In ammonia-rich medium, GDH was the predominant pathway of ammonia assimilation. In nitrogen-fixing
Nitrogen Assimilation by Bacillus licheniformis Organisms Growing in Chemostat Cultures
Meers, J. and L. Pederson
Microbiology, 70, 277-286 (1972)
R T Briggs et al.
The Journal of cell biology, 67(3), 566-586 (1975-12-01)
The ultrastructural localization of NADH oxidase, a possible enzyme in the increased oxidative activity of polymorphonuclear leukocytes (PMN) during phagocytosis, was studied. A new cytochemical technique for the localization of H2O2, a product of NADH oxidase activity, was developed. Cerous
Y Nishiyama et al.
Journal of bacteriology, 183(8), 2431-2438 (2001-03-29)
Amphibacillus xylanus and Sporolactobacillus inulinus NADH oxidases belonging to the peroxiredoxin oxidoreductase family show extremely high peroxide reductase activity for hydrogen peroxide and alkyl hydroperoxides in the presence of the small disulfide redox protein, AhpC (peroxiredoxin). In order to investigate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service